Tetrabutylammonium benzoate 98%

Supplier: Ambeed
Warning

A680768-250MG A680768-1G A680768-5G
76790-974EA 41.38 USD
76790-974 77573-208 77573-210
Tetrabutylammonium benzoate 98%
Tetrabutylammonium benzoate

Precaución: for Research Use Only

Formula: C₂₃H₄₂NO₂
MW: 363.58 g/mol
MDL Number: MFCD00171701
CAS Number: 18819-89-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR