(R)-4-Boc-2-(aminomethyl)morpholine 97%

Supplier: Ambeed
Warning

A680531-5G A680531-1G
76790-972EA 219 USD
76790-972 77569-296
(R)-4-Boc-2-(aminomethyl)morpholine 97%
(R)-4-Boc-2-(aminomethyl)morpholine

Precaución: For research use only.

Formula: C₁₀H₂₀N₂O₃
MDL Number: MFCD13189480
CAS Number: 1174913-80-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR