Tranilast 99+%

Supplier: Ambeed
Warning

Synonyms: 2-[[3-(3,4-Dimethoxyphenyl)-1-oxo-2-propenyl]amino]benzoic Acid, 2-(3-(3,4-Dimethoxyphenyl)acrylamido)benzoic acid, N-(3,4-Dimethoxycinnamoyl)anthranilic Acid

A675778-100MG A675778-1G A675778-250MG A675778-25G A675778-50MG A675778-5G
76790-952EA 6.37 USD
76790-952 76790-954 76790-956 76790-958 76790-960 76790-962
Tranilast 99+%
Tranilast

Precaución: for Research Use Only

Formula: C₁₈H₁₇NO₅
MW: 327.34 g/mol
MDL Number: MFCD00864787
CAS Number: 53902-12-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR