(3R,4R)-tert-Butyl-3-amino-4-hydroxypyrrolidine-1-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: (3R,4R)-tert-Butyl 3-amino-4-hydroxypyrrolidine-1-carboxylate, trans-3-Amino-4-hydroxypyrrolidine, N1-BOC protected

A673464-100MG A673464-1G A673464-250MG A673464-5G
76790-944EA 40.32 USD
76790-944 76790-946 76790-948 76790-950
(3R,4R)-tert-Butyl-3-amino-4-hydroxypyrrolidine-1-carboxylate 98%
(3R,4R)-tert-Butyl-3-amino-4-hydroxypyrrolidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₉H₁₈N₂O₃
MW: 202.26 g/mol
MDL Number: MFCD11111251
CAS Number: 330681-18-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR