4-(Chloromethyl)styrene 90% stabilized

Supplier: Ambeed
Danger

Synonyms: 1-(Chloromethyl)-4-vinylbenzene, 4-Vinylbenzyl Chloride

Stabilization: Stabilised with 4-tert-butylpyrocatechol (TBC)

A672315-5G A672315-25G
76790-936EA 11.67 USD
76790-936 77595-024
4-(Chloromethyl)styrene 90% stabilized
4-(Chloromethyl)styrene

Precaución: for Research Use Only

Formula: C₉H₉Cl
MW: 152.62 g/mol
MDL Number: MFCD00051362
CAS Number: 1592-20-7
UN: 3265
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR