Torsemide 99%

Supplier: Ambeed
Warning

Synonyms: Torasemide, N-(Isopropylcarbamoyl)-4-(m-tolylamino)pyridine-3-sulphonamide

A671193-100MG A671193-10MG A671193-1G
76790-926EA 6.37 USD
76790-926 76790-928 76844-696
Torsemide 99%
Torsemide

Precaución: for Research Use Only

Formula: C₁₆H₂₀N₄O₃S
MW: 348.42 g/mol
MDL Number: MFCD00866166
CAS Number: 56211-40-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR