Diethyl (2-phenylethyl)malonate 97%

Supplier: Ambeed
Warning

Synonyms: Diethyl 2-phenethylmalonate

A670733-100G A670733-1G A670733-25G A670733-5G
76790-918EA 501.81 USD
76790-918 76790-920 76790-922 76790-924
Diethyl (2-phenylethyl)malonate 97%
Diethyl (2-phenylethyl)malonate

Precaución: for Research Use Only

Formula: C₁₅H₂₀O₄
MW: 264.32 g/mol
MDL Number: MFCD00026887
CAS Number: 6628-68-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR