(2-Acetamidophenyl)boronic acid pinacol ester 96%

Supplier: Ambeed
Warning

Synonyms: 2-Acetamidobenzeneboronic acid pinacol ester, N-(2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)acetamide, 2-Acetylaminophenylboronic acid, pinacol ester

A669136-100MG A669136-1G A669136-250MG A669136-5G
76790-908EA 14 USD
76790-908 76790-910 76790-912 77873-071
(2-Acetamidophenyl)boronic acid pinacol ester 96%
(2-Acetamidophenyl)boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₂₀BNO₃
MDL Number: MFCD02179480
CAS Number: 380430-61-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR