Tirofiban hydrochloride monohydrate 98%

Supplier: Ambeed
Warning

Synonyms: N-(Butylsulfonyl)-O-[4-(4-piperidinyl)butyl]-L-tyrosine hydrochloride monohydrate, MK-383 HCl, (S)-2-(Butylsulphonamido)-3-(4-(4-(piperidin-4-yl)butoxy)phenyl)propanoic acid hydrochloride hydrate

A662355-10MG A662355-1G A662355-1MG A662355-25MG A662355-50MG A662355-5MG A662355-250MG A662355-5G
76790-892EA 9 USD
76790-892 76790-894 76790-896 76790-898 76790-900 76790-902 77602-686 77602-688
Tirofiban hydrochloride monohydrate 98%
Tirofiban hydrochloride monohydrate

Precaución: for Research Use Only

Formula: C₂₂H₃₉ClN₂O₆S
MDL Number: MFCD07368623
CAS Number: 150915-40-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR