Pivalonitrile 99%

Supplier: Ambeed
Danger

A660022-25G A660022-5G
76790-886EA 8 USD
76790-886 76790-888
Pivalonitrile 99%
Pivalonitrile

Precaución: for Research Use Only

Formula: C₅H₉N
MDL Number: MFCD00001847
CAS Number: 630-18-2
UN: 1993
ADR: 3,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR