1-(tert-Butyldimethylsilyl)imidazole 95%

Supplier: Ambeed
Warning

Synonyms: 1-(tert-Butyldimethylsilyl)-1H-imidazole

A659091-1G A659091-5G
76790-880EA 36.08 USD
76790-880 76790-882
1-(tert-Butyldimethylsilyl)imidazole 95%
1-(tert-Butyldimethylsilyl)imidazole

Precaución: for Research Use Only

Formula: C₉H₁₈N₂Si
MW: 182.34 g/mol
MDL Number: MFCD00011682
CAS Number: 54925-64-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR