Udp-alpha-d-n-acetylgalactosamine,disodiumsalt 98%

Supplier: Ambeed
Warning

Synonyms: UDP-Alpha-d-n-acetylgalactosamine, disodium salt

A658657-10MG A658657-25MG A658657-100MG
76790-876EA 94.43 USD
76790-876 76790-878 77602-680
Udp-alpha-d-n-acetylgalactosamine,disodiumsalt 98%
Udp-alpha-d-n-acetylgalactosamine,disodiumsalt

Precaución: for Research Use Only

Formula: C₁₇H₂₅N₃Na₂O₁₇P₂
MW: 651.31 g/mol
MDL Number: MFCD00077894
CAS Number: 108320-87-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR