Bis(8-quinolinolato)zinc(II) hydrate 98+%

Supplier: Ambeed
Warning

Synonyms: Zinc 8-hydroxyquinolinate, Bis(8-hydroxyquinolinato)zinc(II), Zinc(II) Bis(8-hydroxyquinolinate)

A656441-25G A656441-5G
76790-866EA 173 USD
76790-866 76790-868
Bis(8-quinolinolato)zinc(II) hydrate 98+%
Bis(8-quinolinolato)zinc(II) hydrate

Precaución: for Research Use Only

Formula: C₁₈H₁₂N₂O₂Zn
MW: 353.69 g/mol
MDL Number: MFCD00065291
CAS Number: 13978-85-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR