tert-Butyl-4-methoxycarbanilate 95%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl N-(4-methoxyphenyl)carbamate, tert-Butyl (4-methoxyphenyl)carbamate

A656099-100G A656099-1G A656099-25G A656099-5G
76790-858EA 315.09 USD
76790-858 76790-860 76790-862 76790-864
tert-Butyl-4-methoxycarbanilate 95%
tert-Butyl-4-methoxycarbanilate

Precaución: for Research Use Only

Formula: C₁₂H₁₇NO₃
MW: 223.268 g/mol
MDL Number: MFCD00816766
CAS Number: 18437-68-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR