Z-Leu-OMe 98%

Supplier: Ambeed
Warning

A655061-25G A655061-5G
76790-854EA 45 USD
76790-854 76790-856
Z-Leu-OMe 98%
Z-Leu-OMe

Precaución: for Research Use Only

Formula: C₁₅H₂₁NO₄
MDL Number: MFCD03424106
CAS Number: 51021-87-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR