9-Ethyl-9H-carbazol-2-amine 95+%

Supplier: Ambeed
Danger

A648032-1G A648032-250MG
76790-848EA 715.05 USD
76790-848 76790-850
9-Ethyl-9H-carbazol-2-amine 95+%
9-Ethyl-9H-carbazol-2-amine

Precaución: for Research Use Only

Formula: C₁₄H₁₄N₂
MW: 210.27 g/mol
MDL Number: MFCD00004964
CAS Number: 40444-36-8
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR