5-Methoxypyrimidine 97%

Supplier: Ambeed
Danger

A641187-100MG A641187-250MG A641187-5G
76790-816EA 27.59 USD
76790-816 76790-818 76790-820
5-Methoxypyrimidine 97%
5-Methoxypyrimidine

Precaución: for Research Use Only

Formula: C5H6N2O
MW: 110.1100 g/mol
MDL Number: MFCD04971313
CAS Number: 31458-33-0
UN: 1325
ADR: 4.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR