1-Bromo-4-(difluoromethyl)benzene 95%

Supplier: Ambeed
Danger

Synonyms: 4-Bromobenzal fluoride

A637081-10G A637081-1G A637081-250MG A637081-5G
76790-804EA 130.5 USD
76790-804 76790-806 76790-808 76790-810
1-Bromo-4-(difluoromethyl)benzene 95%
1-Bromo-4-(difluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₇H₅BrF₂
MW: 207.02 g/mol
MDL Number: MFCD06657964
CAS Number: 51776-71-7
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR