tert-Butyl 8-hydroxy-3,4-dihydroquinoline-1(2H)-carboxylate 95%
Supplier: Ambeed
|
Warning
|
A634083-100MG
A634083-1G
A634083-250MG
76790-796EA
31.83
USD
76790-796
76790-798
76790-800
tert-Butyl 8-hydroxy-3,4-dihydroquinoline-1(2H)-carboxylate 95%
tert-Butyl 8-hydroxy-3,4-dihydroquinoline-1(2H)-carboxylate
Precaución: for Research Use Only
Formula:
C₁₄H₁₉NO₃ MW: 249.30 g/mol |
MDL Number:
MFCD08275030 CAS Number: 676255-10-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...