3-(Bromomethyl)-1H-indole, N-BOC protected 95%

Supplier: Ambeed
Danger

Synonyms: tert-Butyl 3-(bromomethyl)-1H-indole-1-carboxylate, 3-(Bromomethyl)-1-(tert-butoxycarbonyl)-1H-indole

A632931-1G A632931-250MG A632931-5G
76790-790EA 61.54 USD
76790-790 76790-792 76790-794
3-(Bromomethyl)-1H-indole, N-BOC protected 95%
3-(Bromomethyl)-1H-indole, N-BOC protected

Precaución: for Research Use Only

Formula: C₁₄H₁₆BrNO₂
MW: 310.186 g/mol
MDL Number: MFCD05864361
CAS Number: 96551-21-2
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR