5-Bromo-4-chloro-2-fluoropyridine 98%

Supplier: Ambeed
Warning

A632843-100MG A632843-1G A632843-250MG A632843-5G
76790-784EA 30 USD
76790-784 76790-786 76790-788 77573-052
5-Bromo-4-chloro-2-fluoropyridine 98%
5-Bromo-4-chloro-2-fluoropyridine

Precaución: for Research Use Only

Formula: C₅H₂BrClFN
MDL Number: MFCD12911587
CAS Number: 1184920-15-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR