Tris(4-methylphenyl)phosphine oxide 98%

Supplier: Ambeed
Danger

A632712-1G A632712-250MG A632712-25G A632712-5G
76790-776EA 24 USD
76790-776 76790-778 76790-780 76790-782
Tris(4-methylphenyl)phosphine oxide 98%
Tris(4-methylphenyl)phosphine oxide

Precaución: for Research Use Only

Formula: C₂₁H₂₁OP
MDL Number: MFCD00059321
CAS Number: 797-70-6
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR