Tetramethylammonium tetrafluoroborate 99%

Supplier: Ambeed
Danger

Synonyms: Tetramethylammoniumtetrafluoroborate

A629423-1G A629423-25G A629423-5G
76790-752EA 5.31 USD
76790-752 76790-754 76790-756
Tetramethylammonium tetrafluoroborate 99%
Tetramethylammonium tetrafluoroborate

Precaución: for Research Use Only

Formula: C₄H₁₂BF₄N
MW: 160.95 g/mol
MDL Number: MFCD00011745
CAS Number: 661-36-9
UN: 1759
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR