2-Bromo-4,5-difluorobenzonitrile 97%

Supplier: Ambeed
Warning

A629121-100G A629121-10G A629121-1G A629121-25G A629121-5G
76790-742EA 179 USD
76790-742 76790-744 76790-746 76790-748 76790-750
2-Bromo-4,5-difluorobenzonitrile 97%
2-Bromo-4,5-difluorobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₂BrF₂N
MDL Number: MFCD09864661
CAS Number: 64695-82-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR