Vitamin K1 (phytomenadione, phylloquinone) 98%

Supplier: Ambeed
Warning

Synonyms: 2-Methyl-3-((7R,11R,E)-3,7,11,15-tetramethylhexadec-2-en-1-yl)naphthalene-1,4-dione

A627922-100G A627922-1G A627922-250MG A627922-25G A627922-5G
76790-726EA 604.72 USD
76790-726 76790-728 76790-730 76790-732 76790-734
Vitamin K1 (phytomenadione, phylloquinone) 98%
Vitamin K1 (phytomenadione, phylloquinone)

Precaución: for Research Use Only

Formula: C₃₁H₄₆O₂
MW: 450.6958 g/mol
MDL Number: MFCD00214063
CAS Number: 84-80-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR