(4-Phenoxyphenyl)acetic acid 96%

Supplier: Ambeed
Warning

Synonyms: 4-Phenoxyphenylacetic Acid

A626293-100G A626293-10G A626293-1G A626293-25G A626293-5G
76790-706EA 642.91 USD
76790-706 76790-708 76790-710 76790-712 76790-714
(4-Phenoxyphenyl)acetic acid 96%
(4-Phenoxyphenyl)acetic acid

Precaución: for Research Use Only

Formula: (C₆H₅OC₆H₅)CH₂COOH
MW: 228.25 g/mol
MDL Number: MFCD00079779
CAS Number: 6328-74-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR