Diethyl-2,5-dioxobicyclo[2.2.2]octane-1,4-dicarboxylate 95%

Supplier: Ambeed
Warning

A626145-100MG A626145-250MG
76790-702EA 15 USD
76790-702 76790-704
Diethyl-2,5-dioxobicyclo[2.2.2]octane-1,4-dicarboxylate 95%
Diethyl-2,5-dioxobicyclo[2.2.2]octane-1,4-dicarboxylate

Precaución: for Research Use Only

Formula: C₁₄H₁₈O₆
MDL Number: MFCD00213759
CAS Number: 843-59-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR