Tetramethylammonium hydrogen sulphate 96%

Supplier: Ambeed
Warning

Synonyms: Tetramethylammonium hydrogensulphate, Tetramethylammonium bisulphate

A623412-100G A623412-25G A623412-5G A623412-500G
76790-688EA 158 USD
76790-688 76790-690 76790-692 77873-019
Tetramethylammonium hydrogen sulphate 96%
Tetramethylammonium hydrogen sulphate

Precaución: for Research Use Only

Formula: C₄H₁₃NO₄S
MW: 171.22 g/mol
MDL Number: MFCD00036149
CAS Number: 80526-82-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR