2-(2-(2-Amino-6-chloro-9H-purin-9-yl)ethyl)propane-1,3-diyl diacetate 95%

Supplier: Ambeed
Warning

A622851-100G A622851-10G A622851-1G A622851-25G A622851-5G
76790-672EA 194.15 USD
76790-672 76790-674 76790-676 76790-678 76790-680
2-(2-(2-Amino-6-chloro-9H-purin-9-yl)ethyl)propane-1,3-diyl diacetate 95%
2-(2-(2-Amino-6-chloro-9H-purin-9-yl)ethyl)propane-1,3-diyl diacetate

Precaución: for Research Use Only

Formula: C₁₄H₁₈ClN₅O₄
MW: 355.77 g/mol
MDL Number: MFCD08458206
CAS Number: 97845-60-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR