Tin(IV) oxide 99%
Supplier: Ambeed
|
Warning
|
Synonyms:
Tin(iv)oxide, Stannic Oxide, Tin dioxide
A621734-500G
A621734-25G
76790-670EA
74
USD
76790-670
77669-678
Tin(IV) oxide 99%
Tin(IV) oxide
Precaución: for Research Use Only
Formula:
O₂Sn MW: 150.71 g/mol Melting Pt: 1624.85 |
MDL Number:
MFCD00011244 CAS Number: 18282-10-5 Merck Index: 14,08778 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...