2-Chloro-6-fluorophenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (2-Chloro-6-fluorophenyl)boronic acid, (2-Chloro-6-fluorophenyl)metaboric acid, 2-Chloro-6-fluorobenzeneboronic acid, Dihydroxy(2-chloro-6-fluorophenyl)borane

A616981-1G A616981-5G A616981-25G A616981-100G A616981-10G
76790-646EA 6.37 USD
76790-646 76790-648 77602-554 77669-652 77669-654
2-Chloro-6-fluorophenylboronic acid 98%
2-Chloro-6-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₅BClFO₂
MW: 174.37 g/mol
MDL Number: MFCD04039892
CAS Number: 313545-32-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR