Voriconazole ≥99%

Supplier: Ambeed
Danger

A615493-100MG A615493-1MG A615493-250MG A615493-50MG A615493-1G A615493-10MG A615493-5G A615493-25G
76790-636EA 37 USD
76790-636 76790-638 76790-640 76790-642 77551-846 77669-644 77669-646 77873-011
Voriconazole ≥99%
Voriconazole

Precaución: for Research Use Only

Formula: C₁₆H₁₄F₃N₅O
MDL Number: MFCD00905717
CAS Number: 137234-62-9
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR