(S)-(2'-Isopropoxy-[1,1'-binaphthalen]-2-yl)diphenylphosphine 98%

Supplier: Ambeed
Warning

A609123-100MG A609123-1G A609123-250MG
76790-616EA 90 USD
76790-616 76790-618 76790-620
(S)-(2'-Isopropoxy-[1,1'-binaphthalen]-2-yl)diphenylphosphine 98%
(S)-(2'-Isopropoxy-[1,1'-binaphthalen]-2-yl)diphenylphosphine

Precaución: for Research Use Only

Formula: C₃₅H₂₉OP
CAS Number: 137769-30-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR