9-Vinylcarbazole 98% stabilized

Supplier: Ambeed
Danger


Stabilization: Stabilised with 4-tert-butylpyrocatechol (TBC)

A607777-10G A607777-5G
76790-612EA 12.74 USD
76790-612 76790-614
9-Vinylcarbazole 98% stabilized
9-Vinylcarbazole

Precaución: for Research Use Only

Formula: C₁₄H₁₁N
MW: 193.25 g/mol
MDL Number: MFCD00004966
CAS Number: 1484-13-5
UN: 2811
ADR: 6.1,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR