5-Methyl-2-phenyl-1,2-dihydropyrazol-3-one 98+%

Supplier: Ambeed
Warning

A607737-100G A607737-25G A607737-500G A607737-5G
76790-604EA 98 USD
76790-604 76790-606 76790-608 76790-610
5-Methyl-2-phenyl-1,2-dihydropyrazol-3-one 98+%
5-Methyl-2-phenyl-1,2-dihydropyrazol-3-one

Precaución: for Research Use Only

Formula: C10H10N2O
MDL Number: MFCD00127930
CAS Number: 19735-89-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR