Disodium succinate 99%

Supplier: Ambeed
Warning

Synonyms: 1,4-Butanedioic acid disodium salt, Sodium Succinate, Succinic acid disodium salt, Sodium succinate dibasic

A605375-100G A605375-25G A605375-1000G A605375-500G
76790-600EA 9 USD
76790-600 76790-602 77572-956 77602-532
Disodium succinate 99%
Disodium succinate

Precaución: for Research Use Only

Formula: C₄H₄Na₂O₄
MW: 162.05 g/mol
MDL Number: MFCD00002790
CAS Number: 150-90-3
Merck Index: 12,08828

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR