Tris(4-aminophenyl)methane 97%

Supplier: Ambeed
Warning

Synonyms: Leucopararosaniline, 4,4',4''-Methanetriyltrianiline

A601577-100MG A601577-1G A601577-1MG A601577-250MG A601577-25G A601577-5G
76790-562EA 35.01 USD
76790-562 76790-564 76790-566 76790-568 76790-570 76790-572
Tris(4-aminophenyl)methane 97%
Tris(4-aminophenyl)methane

Precaución: for Research Use Only

Formula: C₁₉H₁₉N₃
MW: 289.38 g/mol
MDL Number: MFCD00137820
CAS Number: 548-61-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR