1-((1-((1-Methoxypropan-2-yl)oxy)propan-2-yl)oxy)propan-2-ol 98%

Supplier: Ambeed
Warning

A600805-1000G A600805-100G A600805-500G A600805-25G
76790-556EA 54.11 USD
76790-556 76790-558 76790-560 77602-508
1-((1-((1-Methoxypropan-2-yl)oxy)propan-2-yl)oxy)propan-2-ol 98%
1-((1-((1-Methoxypropan-2-yl)oxy)propan-2-yl)oxy)propan-2-ol

Precaución: for Research Use Only

Formula: C₁₀H₂₂O₄
MW: 206.27 g/mol
MDL Number: MFCD00059142
CAS Number: 20324-33-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR