Tetraammineplatinum(II) chloride monohydrate 98%

Supplier: Ambeed
Warning

Synonyms: Tetraammineplatinum (II) chloride hydrate

A600421-1G A600421-250MG A600421-5G
76790-550EA 127.31 USD
76790-550 76790-552 76790-554
Tetraammineplatinum(II) chloride monohydrate 98%
Tetraammineplatinum(II) chloride monohydrate

Precaución: for Research Use Only

Formula: Cl₂H₁₄N₄OPt
MW: 352.123 g/mol
MDL Number: MFCD00149947
CAS Number: 13933-33-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR