2-tert-Butylhydroquinone 99+%

Supplier: Ambeed
Warning

Synonyms: t-Butylhydroquinone, 2-t-Butylhydroquinone, 2-(tert-Butyl)benzene-1,4-diol(may occur to produce black solid), tert-Butylhydroquinone, TBHQ

A598680-1000G A598680-100G A598680-25G A598680-500G A598680-5G
76790-538EA 71.09 USD
76790-538 76790-540 76790-542 76790-544 76790-546
2-tert-Butylhydroquinone 99+%
2-tert-Butylhydroquinone

Precaución: for Research Use Only

Formula: C₁₀H₁₄O₂
MW: 166.22 g/mol
MDL Number: MFCD00002344
CAS Number: 1948-33-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR