1-Methoxy-2,2,2-trifluoroethanol 90%

Supplier: Ambeed
Danger

Synonyms: Trifluoroacetaldehyde methyl hemiacetal

A596394-5G A596394-1G
76790-528EA 39.26 USD
76790-528 77602-484
1-Methoxy-2,2,2-trifluoroethanol 90%
1-Methoxy-2,2,2-trifluoroethanol

Precaución: for Research Use Only

Formula: C₃H₅F₃O₂
MW: 130.07 g/mol
MDL Number: MFCD00013572
CAS Number: 431-46-9
UN: 3271
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR