4'-Propyl-4-biphenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (4'-Propyl-[1,1'-biphenyl]-4-yl)boronic acid

A596152-1G A596152-25G A596152-5G
76790-522EA 10.61 USD
76790-522 76790-524 76790-526
4'-Propyl-4-biphenylboronic acid 98%
4'-Propyl-4-biphenylboronic acid

Precaución: for Research Use Only

Formula: C₁₅H₁₇BO₂
MW: 240.11 g/mol
MDL Number: MFCD01114670
CAS Number: 153035-56-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR