Phenethyl-p-toluenesulfonate 98%

Supplier: Ambeed
Warning

Synonyms: Phenethyl 4-methylbenzenesulphonate, 2-Phenylethyl p-Toluenesulphonate, p-Toluenesulphonic acid Phenethyl Ester, p-Toluenesulphonic acid 2-Phenylethyl Ester

A595076-1G A595076-5G
76790-518EA 7.43 USD
76790-518 76790-520
Phenethyl-p-toluenesulfonate 98%
Phenethyl-p-toluenesulfonate

Precaución: for Research Use Only

Formula: C₁₅H₁₆O₃S
MW: 276.35 g/mol
MDL Number: MFCD00059322
CAS Number: 4455-09-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR