Methyl-1-benzofuran-2-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl benzofuran-2-carboxylate

A593895-1G A593895-250MG
76790-510EA 92.3 USD
76790-510 76790-512
Methyl-1-benzofuran-2-carboxylate 98%
Methyl-1-benzofuran-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₈O₃
MW: 176.17 g/mol
MDL Number: MFCD01159618
CAS Number: 1646-27-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR