Tetrapropylammonium hydroxide 40% (w/w) in water

Supplier: Ambeed
Danger

A592487-25G A592487-100G
76790-504EA 6 USD
76790-504 77602-478
Tetrapropylammonium hydroxide 40% (w/w) in water
Tetrapropylammonium hydroxide

Precaución: for Research Use Only

Formula: C₁₂H₂₉NO
MDL Number: MFCD00009360
CAS Number: 4499-86-9
UN: 3267
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR