Sulfacarbamide 98%

Supplier: Ambeed
Warning

A591868-1MG A591868-25MG
76790-500EA 6 USD
76790-500 76790-502
Sulfacarbamide 98%
Sulfacarbamide

Precaución: for Research Use Only

Formula: C₇H₉N₃O₃S
MDL Number: MFCD00025437
CAS Number: 547-44-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR