Tetrabutylammonium toluene-4-sulfonate 98%

Supplier: Ambeed
Warning

Synonyms: Tetrabutylammonium p-Toluenesulphonate, Tetrabutylammonium 4-methylbenzenesulphonate

A590695-100G A590695-25G A590695-500G
76790-476EA 92.3 USD
76790-476 76790-478 76790-480
Tetrabutylammonium toluene-4-sulfonate 98%
Tetrabutylammonium toluene-4-sulphonate

Precaución: for Research Use Only

Formula: C₂₃H₄₃NO₃S
MW: 413.6574 g/mol
MDL Number: MFCD00043227
CAS Number: 7182-86-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR