tert-Butyl-2,7-diazaspiro[4.5]decane-2-carboxylate ≥97%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 2,7-diazaspiro[4.5]decane-2-carboxylate

A590403-100MG A590403-1G A590403-250MG
76790-470EA 68.96 USD
76790-470 76790-472 76790-474
tert-Butyl-2,7-diazaspiro[4.5]decane-2-carboxylate ≥97%
tert-Butyl-2,7-diazaspiro[4.5]decane-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₃H₂₄N₂O₂
MW: 240.35 g/mol
MDL Number: MFCD08234437
CAS Number: 885268-42-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR