3-Amino-1-ethyl-1H-pyrazole 98%

Supplier: Ambeed
Danger

A588198-1G A588198-250MG
76790-460EA 65 USD
76790-460 76790-462
3-Amino-1-ethyl-1H-pyrazole 98%
3-Amino-1-ethyl-1H-pyrazole

Precaución: for Research Use Only

Formula: C₅H₉N₃
MDL Number: MFCD03422571
CAS Number: 55361-49-4
UN: 2735
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR