4-Acetoxymethylbenzeneboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (4-(Acetoxymethyl)phenyl)boronic acid

A588061-1G A588061-250MG A588061-5G
76790-454EA 101 USD
76790-454 76790-456 76790-458
4-Acetoxymethylbenzeneboronic acid 98%
4-Acetoxymethylbenzeneboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₁BO₄
MDL Number: MFCD08235082
CAS Number: 326496-51-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR